Learn More
Electron Microscopy Sciences Power Max Cutters, 6.5"
SDP
Catalog No. 5030645
Encompass
Ergonomic compound cutters that offer the finest in compound leverage construction at an affordable price. Leverage combined with induction-hardened, high-grade steel jaws allows for extra close, flush cutting. The handles are shaped to be ergonomically and comfortably.
OAL: 6.5"
Catalog No. 50-306-45 Supplier Electron Microscopy Sciences Supplier No. 6218710
Ergonomic compound cutters that offer the finest in compound leverage construction at an affordable price. Leverage combined with induction-hardened, high-grade steel jaws allows for extra close, flush cutting. The handles are shaped to be ergonomically and comfortably.
OAL: 6.5"