Learn More
Bio Rad Laboratories POWER PAC H.C.
Catalog No. NC9069331
Encompass
PowerPac HC Power Supply Input Power (nominal): 100 to 120/220 to 240VAC, Autoswitching, Output Specifications: 250V, 3.0A, 300w, Output Range: (programmable): 5 to 250V, Fully adjustable in 1V increments, Dimensions: 25 x 28.5 x 8cm.
Catalog No. NC9069331 Supplier Bio Rad Laboratories Supplier No. 1645052
PowerPac HC Power Supply Input Power (nominal): 100 to 120/220 to 240VAC, Autoswitching, Output Specifications: 250V, 3.0A, 300w, Output Range: (programmable): 5 to 250V, Fully adjustable in 1V increments, Dimensions: 25 x 28.5 x 8cm.