Learn More
RS AMERICAS INC POWER SONIC PS-1270-F2
Catalog No. NC0847778
Encompass
Battery; Sealed Lead Acid; Rechargeable; 12 V; 7 Ah; 4.80 Lbs.; Spill-Proof; F2 Quick Tabs. Absorbent Glass Mat (Agm) Technology For Superior Performance. Valve Regulated, Spill Proof Construction Allows Safe Operation In Any Position. Power/Volume Ratio Yielding Unrivaled Energy Density. Rugged Impact Resistant Abs Case And Cover (Ul94-Hb). Approved For Transport By Air. D.O.T., I.A.T.A., F.A.A. And C.A.B. Certified. U.L. Recognized Under Fi Le Number Mh 20845.
Catalog No. NC0847778 Supplier RS AMERICAS INC Supplier No. 70115580
Battery; Sealed Lead Acid; Rechargeable; 12 V; 7 Ah; 4.80 Lbs.; Spill-Proof; F2 Quick Tabs. Absorbent Glass Mat (Agm) Technology For Superior Performance. Valve Regulated, Spill Proof Construction Allows Safe Operation In Any Position. Power/Volume Ratio Yielding Unrivaled Energy Density. Rugged Impact Resistant Abs Case And Cover (Ul94-Hb). Approved For Transport By Air. D.O.T., I.A.T.A., F.A.A. And C.A.B. Certified. U.L. Recognized Under Fi Le Number Mh 20845.