Learn More
Ace Glass, Inc. 450 watt UV mercury-arc lamp power supply, input 120v 60Hz, USA model, not NRTL certified
SDP
Catalog No. NC0387984
Encompass
450watt cased UV power supply that supplies the extra voltage and current required to initiate the lamps arc and reduce operational power. For use with Hanovia medium-pressure UV immersion lamps and reflectors. Input: 120v 60Hz.
Catalog No. NC0387984 Supplier Ace Glass, Inc. Supplier No. 783060
450watt cased UV power supply that supplies the extra voltage and current required to initiate the lamps arc and reduce operational power. For use with Hanovia medium-pressure UV immersion lamps and reflectors. Input: 120v 60Hz.