Learn More
Grainger Power Strip 60 W x 2 D x 2 in H White
Catalog No. 502767632
Encompass_Preferred
The Grainger Approved 6-Outlet Power Strip Surge Protector provides reliable power distribution and surge protection for computers, printers, and other sensitive electronics. This 15-foot power strip has a flat plug that allows furniture to be placed flush against the wall. It features a 15-amp circuit breaker, 6 outlets, an illuminated on/off switch, and a 6-foot 14-gauge power cord. The power strip's all-metal housing protects against EMI/RFI line noise. It has a peak let-through voltage of 330V and conforms to UL 1449 3rd Edition safety standards. This is an affordable option for protecting equipment from power surges and spikes.
Catalog No. 50-276-7632 Supplier Grainger Supplier No. 16Y925
The Grainger Approved 6-Outlet Power Strip Surge Protector provides reliable power distribution and surge protection for computers, printers, and other sensitive electronics. This 15-foot power strip has a flat plug that allows furniture to be placed flush against the wall. It features a 15-amp circuit breaker, 6 outlets, an illuminated on/off switch, and a 6-foot 14-gauge power cord. The power strip's all-metal housing protects against EMI/RFI line noise. It has a peak let-through voltage of 330V and conforms to UL 1449 3rd Edition safety standards. This is an affordable option for protecting equipment from power surges and spikes.