Learn More
Poltex LLC Power Strip Holder (3 Magnets)
Description
- Magnet mounted power strip holder with cut-outs and slots for zip tie, or other tie down to secure power strip
- Attach to metal surface with powerful magnets (included)
- Durable and cleanable PETG construction
- Power strip not included
Specifications
Specifications
| Product Type | Power Strip Holder |
| For Use With (Equipment) | Power Strip |
| Length (English) | 12 in. |
| Material | PETG |
| Autoclavable | Not Autoclavable |