Learn More
Hoefer, Inc Hoefer™ PS200HC 200 Volt High Current Power Supply
SDP
Catalog No. NC2741114
Encompass_Preferred
The PS200HC is ideal for large electrophoretic blotting applications and for low voltage electrophoresis runsFeatures:Constant voltage or constant current mode with automatic crossover5 to 200 V, 10 to 2000 mA, 200 WFour sets of 4 mm output jacksIdeal choice for blotting applicationsContinuous runs or 999 minute timerAutomatic recovery after power failureOpen circuit detectionShort circuit protectionModes of Operation:Constant Voltage Mode—the power supply will attempt to maintain the user selected voltage by automatically adjusting the current flowConstant Current Mode—the power supply will attempt to maintain the user selected current flow by automatically adjusting the voltage suppliedAutomatic Crossover—when the constant parameter (voltage or current) cannot be maintained due to power supply operating limits the power supply automatically switches to maintaining the other parameter
Catalog No. NC2741114 Supplier Hoefer, Inc Supplier No. PS200HC
Add to Cart
Add to Cart
The PS200HC is ideal for large electrophoretic blotting applications and for low voltage electrophoresis runsFeatures:Constant voltage or constant current mode with automatic crossover5 to 200 V, 10 to 2000 mA, 200 WFour sets of 4 mm output jacksIdeal choice for blotting applicationsContinuous runs or 999 minute timerAutomatic recovery after power failureOpen circuit detectionShort circuit protectionModes of Operation:Constant Voltage Mode—the power supply will attempt to maintain the user selected voltage by automatically adjusting the current flowConstant Current Mode—the power supply will attempt to maintain the user selected current flow by automatically adjusting the voltage suppliedAutomatic Crossover—when the constant parameter (voltage or current) cannot be maintained due to power supply operating limits the power supply automatically switches to maintaining the other parameter