Learn More
Thomas Scientific Enduro Power Supply 300V, 3A, 90W- 115V
SDP
Catalog No. 501459314
Encompass
The Enduro Power Supplies are ideal for DNA, RNA and protein electrophoresis as well as blotting. They provide a high degree of sophistication and versatility yet are extremely easy to use. Two models are available to meet a variety of needs. Dependable performance, intelligent control Simple to program Constant voltage, current or wattage Compact, lightweight and stackable Operating choices for the power supplies include constant voltage, constant amperage or constant wattage (250V model only) and timed or continuous operation. All operating parameters are set using the membrane keypad and displayed digitally on the backlit LCD. Safety features incorporated into the systems include automatic crossover, no load detection, sudden load change and over voltage protection. The units can also be programmed to restart in the event of a power disruption. The Enduro 250V features a voltage range from 5 to 250V in 1V increments. Current is adjustable in 0.01A steps to a maximum of 3A. Wattage i
Catalog No. 50-145-9314 Supplier Thomas Scientific Supplier No. 1222U11
The Enduro Power Supplies are ideal for DNA, RNA and protein electrophoresis as well as blotting. They provide a high degree of sophistication and versatility yet are extremely easy to use. Two models are available to meet a variety of needs. Dependable performance, intelligent control Simple to program Constant voltage, current or wattage Compact, lightweight and stackable Operating choices for the power supplies include constant voltage, constant amperage or constant wattage (250V model only) and timed or continuous operation. All operating parameters are set using the membrane keypad and displayed digitally on the backlit LCD. Safety features incorporated into the systems include automatic crossover, no load detection, sudden load change and over voltage protection. The units can also be programmed to restart in the event of a power disruption. The Enduro 250V features a voltage range from 5 to 250V in 1V increments. Current is adjustable in 0.01A steps to a maximum of 3A. Wattage i