Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson Power Supply

Catalog No. FH07008G Shop All Gilson Products
Change view
Click to view available options
Weight (English):
0.43 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
FH07008G 0.43 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FH07008G Supplier Gilson Supplier No. FH07008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

USB power supply for PIPETMAN™ ensures correct voltage / power input for safe operation

  • The power supply comes with multiple plugs: US, EU, UK, AU - meaning it is designed for international compatibility.
  • The Power Supply for MACROMAN provides the electrical power needed for the MACROMAN pipette controller to operate.
  • Without this supply, the MACROMAN system cannot be powered or used for pipetting

Specifications

Product Type Power Supply
Weight (English) 0.43 lb.
Weight (Metric) 0.2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.