Learn More
Cole-Parmer Argos Technologies FlexiRoll Cell Roller Power Supply Adapter; 100-240 VAC
Catalog No. 501960675 Shop All Cole-Parmer Products
Encompass_Preferred
Compatible for use with the Argos Technologies Flexiroll, models 04397-44 and -44, which are sold separately.
Catalog No. 50-196-0675 Supplier Cole-Parmer Supplier No. 0439747
Compatible for use with the Argos Technologies Flexiroll, models 04397-44 and -44, which are sold separately.