Learn More
Thomas Scientific Power Supply Adapter, 4 to 4mm
SDP
Catalog No. NC1507775
Encompass
1186L41: Extends the length of the standard electrode cables (4mm) of Labnet’s electrophoresis equipment to fit within deeply recessed power supplies. 1186L42: Allows Labnet’s electrophoresis equipment to be run from high voltage power supplies with 2 mm outputs.
Catalog No. NC1507775 Supplier Thomas Scientific Supplier No. 1186L41
Add to Cart
Add to Cart
1186L41: Extends the length of the standard electrode cables (4mm) of Labnet’s electrophoresis equipment to fit within deeply recessed power supplies. 1186L42: Allows Labnet’s electrophoresis equipment to be run from high voltage power supplies with 2 mm outputs.