Learn More
Research Products International Corp Extensions Cables, 48 Inches, One Pair
SDP
Catalog No. 50212277
Encompass
High quality cables for connecting Genie blotters and gel units to D.C. power supplies. Cables supplied in pairs(red/black). Each cable has a one piece molded banana plug on one end and a molded banana jack on other end. Heavy duty 18 gauge flexible wire with durable PVC insulation. Compatible with most electrophoresis power supplies and gel units.
Catalog No. 50-212-277 Supplier Research Products International Corp Supplier No. 144027
High quality cables for connecting Genie blotters and gel units to D.C. power supplies. Cables supplied in pairs(red/black). Each cable has a one piece molded banana plug on one end and a molded banana jack on other end. Heavy duty 18 gauge flexible wire with durable PVC insulation. Compatible with most electrophoresis power supplies and gel units.