Learn More
Cole-Parmer Extech 382200 Single Output Economical DC Power Supply, 30V/1A
Catalog No. 50993009 Shop All Cole-Parmer Products
Encompass
These economical DC power supplies are ideal for bench testing, field service, R & D, educational, and telecommunication applications. Units provide short circuit and overload protection and display both voltage and current on the dual backlit display. Additional features include adjustable constant voltage or constant current output, positive and negative banana plug output jacks, and an illuminated on/off switch.
Catalog No. 50-993-009 Supplier Cole-Parmer Supplier No. 2684975
These economical DC power supplies are ideal for bench testing, field service, R & D, educational, and telecommunication applications. Units provide short circuit and overload protection and display both voltage and current on the dual backlit display. Additional features include adjustable constant voltage or constant current output, positive and negative banana plug output jacks, and an illuminated on/off switch.