Learn More
Cole-Parmer Extech 382213 Digital Regulated DC Power Supply, 115/220 VAC
Catalog No. 50993006 Shop All Cole-Parmer Products
Encompass
Adjustable 0 to 30 V, 0 to 3 A, plus fixed 5 V and 12 V outputs-ideal for almost any application. Current limiting indicator, overload and short circuit protection. View both voltage and current simultaneously on either an analog or a digital display.
Catalog No. 50-993-006 Supplier Cole-Parmer Supplier No. 2684920
Adjustable 0 to 30 V, 0 to 3 A, plus fixed 5 V and 12 V outputs-ideal for almost any application. Current limiting indicator, overload and short circuit protection. View both voltage and current simultaneously on either an analog or a digital display.