Learn More
Fisherbrand™ Power Supply for Fisherbrand™ accumet™ AB and XL Benchtop Meters
Catalog No. 13636104 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters
Catalog No. For Use With (Equipment)
13-636-104 AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters
1 options
Catalog No. 13-636-104
Add to Cart
Add to Cart
Universal replacement power supply
  • For use with accumet benchtop meters models: AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 meters
TRUSTED_SUSTAINABILITY
Specifications
Type Power Supply
Electrical Requirements 110/230 VAC, 9 VDC, 18 W, center positive
For Use With (Equipment) AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters