Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Power Supply for Ripple Tank
GREENER_CHOICESCI_ED

Catalog No. S139445 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
9+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139445 9+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139445 Supplier Supertek™ Supplier No. PH36062
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Supertek™ Power Supply for Ripple Tank PH36044, is to operate both the rippler motor and the illuminate.

  • Output 12 V A.C. max 24 W for illuminate and 0 to 6 V D.C. variable for rippler motor.
  • Operates from 240 V A.C. mains.

Specifications

Grade 9+
Format Model
Languages English
Subject Physical Science - Waves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.