Learn More
LabStrong Corp POWER SWITCH FOR FI-STREEM 2S
Catalog No. NC1023242
Encompass_Preferred
The LabStrong Fi-Streem part number 07260LS compares to Fi-Streem part number 07260. The switch (clear, 240V) is a replacement part for Fi-Streem Water Purification Stills.
Catalog No. NC1023242 Supplier LabStrong Corp Supplier No. 07260LS
The LabStrong Fi-Streem part number 07260LS compares to Fi-Streem part number 07260. The switch (clear, 240V) is a replacement part for Fi-Streem Water Purification Stills.