Learn More
Grainger Power Venter Sidewall 4 In
Catalog No. 502751388
Encompass_Preferred
The Grainger Approved Power Venter Sidewall Fan is a 4 inch diameter sidewall exhaust fan designed for venting bathrooms, laundry rooms and other household areas. It features durable steel construction with a baked enamel finish, thermally protected motor, permanently lubricated sleeve bearing, plug-in power cord, and 4 inch duct connector. This fan has a listed airflow capacity of 70 CFM and operates at 2700 RPM and 0.4 amps. It can mount through walls up to 7 inches thick and includes mounting hardware. This fan meets HVI performance standards for airflow and sound.
Catalog No. 50-275-1388 Supplier Grainger Supplier No. 14U193
The Grainger Approved Power Venter Sidewall Fan is a 4 inch diameter sidewall exhaust fan designed for venting bathrooms, laundry rooms and other household areas. It features durable steel construction with a baked enamel finish, thermally protected motor, permanently lubricated sleeve bearing, plug-in power cord, and 4 inch duct connector. This fan has a listed airflow capacity of 70 CFM and operates at 2700 RPM and 0.4 amps. It can mount through walls up to 7 inches thick and includes mounting hardware. This fan meets HVI performance standards for airflow and sound.