Learn More
MSC Minuteman Cylindrical Scrubber, 28 inch, On Board Charger, Crown Batteries, Nylon Brushes
Catalog No. 502835699
Encompass
This cylindrical scrubber is designed for efficient floor cleaning with a 28 inch cleaning path. It features an on-board charger and is powered by four 275 AH, 6V Crown batteries. Equipped with two cylindrical nylon brushes, it effectively cleans various surfaces. The unit also includes 30 gallon solution and 30 gallon recovery tanks for extended operation.
  • 28 inch cleaning path
  • On-board charger
  • Four 275 AH, 6V batteries
  • Two cylindrical nylon brushes
  • 30 gallon solution and 30 gallon recovery tanks
Catalog No. 50-283-5699 Supplier MSC Supplier No. 43869817
Add to Cart
Add to Cart
Request a Quote
This cylindrical scrubber is designed for efficient floor cleaning with a 28 inch cleaning path. It features an on-board charger and is powered by four 275 AH, 6V Crown batteries. Equipped with two cylindrical nylon brushes, it effectively cleans various surfaces. The unit also includes 30 gallon solution and 30 gallon recovery tanks for extended operation.
  • 28 inch cleaning path
  • On-board charger
  • Four 275 AH, 6V batteries
  • Two cylindrical nylon brushes
  • 30 gallon solution and 30 gallon recovery tanks