Learn More
MSC Minuteman Floor Scrubber, 30 inch Cleaning Path, 30 gallon Tank, Disc Brush, Traction Drive
Catalog No. 502835876
Encompass
This disc brush scrubber features a 30 inch cleaning path and traction drive. Part of the innovative E Series walk-behind scrubber line, this machine can cover 30,000 sq. ft. per hour (2,787 m²). Its low brush deck allows for cleaning around and underneath obstacles, and its compact design enables cleaning in tight spaces. It also features a large tank opening for fast filling.
  • On-board charger allows the machine to be charged anywhere.
  • Brush loading/unloading is easy with a switch.
  • 30 gallon solution and recovery tanks.
  • Squeegees are designed for no operator adjustment.
  • Parabolic squeegee is standard on traction drive models.
  • Easy-to-clean, hygienic recovery tank.
  • Machine is not operable when the on-board charger is plugged in.
  • Adjustable handle and ergonomic hand bails for superior handling.
  • Ergonomic, adjustable speed regulation with a dial.
Catalog No. 50-283-5876 Supplier MSC Supplier No. 45028347
Add to Cart
Add to Cart
Request a Quote
This disc brush scrubber features a 30 inch cleaning path and traction drive. Part of the innovative E Series walk-behind scrubber line, this machine can cover 30,000 sq. ft. per hour (2,787 m²). Its low brush deck allows for cleaning around and underneath obstacles, and its compact design enables cleaning in tight spaces. It also features a large tank opening for fast filling.
  • On-board charger allows the machine to be charged anywhere.
  • Brush loading/unloading is easy with a switch.
  • 30 gallon solution and recovery tanks.
  • Squeegees are designed for no operator adjustment.
  • Parabolic squeegee is standard on traction drive models.
  • Easy-to-clean, hygienic recovery tank.
  • Machine is not operable when the on-board charger is plugged in.
  • Adjustable handle and ergonomic hand bails for superior handling.
  • Ergonomic, adjustable speed regulation with a dial.