Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW™ Sustainability Solutions PowerChem™ PC-045-095GR Green 3.5 mil Neoprene Exam Gloves
SDP

Catalog No. 18999139 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-139 Large
18-999-137 Small
18-999-138 Medium
18-999-140 X-Large
18-999-141 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-999-139 Supplier SW™ Sustainability Solutions Supplier No. K000404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High performance neoprene gloves provide superior resistance to chemicals, punctures, and abrasion.

  • Silicone-free for critical environments
  • Exceeds 1.5 AQL laboratory standards
  • SHA Dermatologically Approved
  • Optimal thickness for durability and tactile sensitivity
  • Latex-free pure neoprene
  • Tear-resistant beaded cuff for easy donning
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory, Healthcare
Color Green
Latex Free Yes
Material Neoprene
Product Type Disposable Gloves
Size Large
Thickness (English) 3 mil
Thickness (Metric) 0.08 mm
EN ISO 374-1 Type C
Glove Cuff Style Beaded
Grip Design Textured Fingers
Model K000404
Length (English) 9.5 in.
Length (Metric) 242 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.