Learn More
Mystaire™ Powercord Access Port
SDP
Catalog No. 15338921 Shop All Mystaire Products
Change view
Click to view available options
For Use With (Equipment):
Mystaire Polycarbonate and Polypropylene Workstations
Catalog No. For Use With (Equipment)
15-338-921 Mystaire Polycarbonate and Polypropylene Workstations
1 options
Catalog No. 15-338-921 Supplier Mystaire™ Supplier No. MYPWP
Add to Cart
Add to Cart
Slip-hatch for use with Mystaire™ polycarbonate and polypropylene workstations.
  • Minimizes eddy current within the ductless fume hood, workstation or enclosure
  • Use with power cord, air hose lines and other tubing associated with laboratory applications
Specifications
For Use With (Equipment) Mystaire Polycarbonate and Polypropylene Workstations
Product Type Access Port