Learn More
MSC Majestic Glove Powercut Mechanics Glove, A9 Cut Level, Medium, 1 Pair
Catalog No. 5000688891
Encompass
These mechanics gloves provide advanced hand protection by incorporating Alycore™ technology for industry-leading resistance against cuts, punctures, and hypodermic needles. Designed for durability and performance, they feature a specialized palm construction suitable for rigorous use in demanding environments.
  • Features eight layers of Alycore™ in the palm and four layers in the fingers
  • Includes durable Armor Skin® palm material and a black knit back
  • Provides ANSI/ISEA 105-2024 cut level A9 protection
  • Achieves ANSI hypodermic needle puncture level 5
  • Designed with a pre-curved finger shape for enhanced ergonomics
  • Equipped with a secure Velcro wrist closure for a custom fit
  • Material is lightweight, flexible, breathable, and washable
Catalog No. 50-006-88891 Supplier MSC Supplier No. 19919273
These mechanics gloves provide advanced hand protection by incorporating Alycore™ technology for industry-leading resistance against cuts, punctures, and hypodermic needles. Designed for durability and performance, they feature a specialized palm construction suitable for rigorous use in demanding environments.
  • Features eight layers of Alycore™ in the palm and four layers in the fingers
  • Includes durable Armor Skin® palm material and a black knit back
  • Provides ANSI/ISEA 105-2024 cut level A9 protection
  • Achieves ANSI hypodermic needle puncture level 5
  • Designed with a pre-curved finger shape for enhanced ergonomics
  • Equipped with a secure Velcro wrist closure for a custom fit
  • Material is lightweight, flexible, breathable, and washable