Learn More
Lonza Walkersville PowerFeed A Bas Powder Kit 10L
Catalog No. 5000496347
Encompass
PowerFeedA Chemically Defined and Protein-free Powder Kit is a non-animal origin powder feed kit containing all compontent to produce 10L of liquid feed The PowerFeed A Chemically Defined and Protein-free Powder Feed is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96347 Supplier Lonza Walkersville Supplier No. BE15044D
PowerFeedA Chemically Defined and Protein-free Powder Kit is a non-animal origin powder feed kit containing all compontent to produce 10L of liquid feed The PowerFeed A Chemically Defined and Protein-free Powder Feed is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS