Learn More
Lonza Walkersville PowerFeed A Basal Powder 100L
Catalog No. 5000496213
Encompass
PowerFeed A Chemically Defined and Protein-free Basal Powder is non-animal origin basal powder which needs addition of ferric citrate (2x BE02-059Q) Powder is enough to produce 100L of liquid The PowerFeed Chemically Defined and Protein-free Basal Powder is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96213 Supplier Lonza Walkersville Supplier No. VPW095J
PowerFeed A Chemically Defined and Protein-free Basal Powder is non-animal origin basal powder which needs addition of ferric citrate (2x BE02-059Q) Powder is enough to produce 100L of liquid The PowerFeed Chemically Defined and Protein-free Basal Powder is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS