Learn More
Lonza Walkersville PowerFeed A Basal Powder 50L
Catalog No. 5000496331
Encompass
PowerFeed A Basal Powder is a chemically defined protein-free and non-animal origin basal powder which needs addition of ferric citrate Powder is sufficient to produce 50L of liquid The feed is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96331 Supplier Lonza Walkersville Supplier No. VPW095F
PowerFeed A Basal Powder is a chemically defined protein-free and non-animal origin basal powder which needs addition of ferric citrate Powder is sufficient to produce 50L of liquid The feed is formulated to maximize cell growth using Lonza CHO cell culture media ProCHO 4 -5 and PowerCHO 2 This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS