Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW Sustainability Solutions PowerForm™ PF-95IN Powder-Free Nitrile Examination Gloves
GREENER_CHOICESDP

Catalog No. 17010345 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-345 Small
17-010-344 X-Large
17-355-292 Medium
17-355-293 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-010-345 Supplier SW Sustainability Solutions Supplier No. N700342
Add to Cart
Add to Cart

Protection that is strong enough to stand up to rigorous use while providing sensitivity and dexterity.

  • Finger textured surface for wet and dry grip
  • Tear-resistant beaded cuff for easy donning
  • Optimal thickness for durability and tactile sensitivity
  • Tested for fentanyl protection to ensure safety in high-risk environments
  • Silicone-free gloves eliminate the risk of contamination in sensitive environments
  • Exceeds 1.5 AQL standards for consistent reliability and reduced defect risk
  • Ergonomic, comfortable fit supports all-day wear with excellent dexterity and tactile sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Chlorinated Yes
Accelerator Free No
Applications Laboratory, Healthcare
AQL 1.5
Color Indigo
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.0 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (English) 4.0 mil
Thickness (Metric) 0.10 mm
Glove Cuff Style Beaded
Model N700342
Series PF-95IN
Length (English) 9.5 in.
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.