Learn More
SW™ Sustainability Solutions PowerForm™ PF-95LG Light Green 5.5 mil Nitrile Exam Gloves - 100 ct
SDP
Catalog No. 18001049 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Small
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-001-049 -
18-001-050 Small
18-001-051 -
18-001-052 -
18-001-053 -
18-001-054 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-001-049 Supplier SW™ Sustainability Solutions Supplier No. N200411
Add to Cart
Add to Cart
Food-handling Light Green gloves help prevent contamination while supporting dexterity and grip.
  • EcoTek™ Sustainable Technology
  • Silicone-Free - Eliminates risk of contamination in sensitive food and industrial processes
  • Exceeds 1.5 AQL Standards - Provides consistent reliability and reduced risk of defects
  • Comfortable Ergonomic Fit - Designed for all-day wear with excellent dexterity and tactile sensitivity
  • Tear-resistant beaded cuff eases donning
  • Finger textured surface for wet and dry grip
  • Tear-resistant beaded cuff for easy donning
  • SHA Dermatologically Approved
TRUSTED_SUSTAINABILITY
EcoTek gloves tested to ASTM D5526-12 Standard Test Method for Determining Anaerobic Biodegradation of Plastic Materials Under Accelerated Landfill Conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated.