Learn More
World Precision Instrument Powerful Ball Joint Rare Earth Magnet
SDP
Catalog No. 502521440
Encompass
Construct holding devices for small parts/equipment
This novel magnetic ball joint has phenomenal holding power for up to 2kg of attached weight wile permitting the ball a full 360° rotation on a 180° axis. You can freely orient your equipment to an infinite number of positions within this rotation. This is made possible by the combination of a steel ball (1mm diameter) and a powerful rare earth magnet contained in the magnet cylinder (10 x 20mm). Convenient M3 attachment sites are provided on both the ball (male) and the magnet base (female).
Catalog No. 50-252-1440 Supplier World Precision Instrument Supplier No. 500871
Construct holding devices for small parts/equipment
This novel magnetic ball joint has phenomenal holding power for up to 2kg of attached weight wile permitting the ball a full 360° rotation on a 180° axis. You can freely orient your equipment to an infinite number of positions within this rotation. This is made possible by the combination of a steel ball (1mm diameter) and a powerful rare earth magnet contained in the magnet cylinder (10 x 20mm). Convenient M3 attachment sites are provided on both the ball (male) and the magnet base (female).