Learn More
Micronova™ PowerHead™ Bar
SDP
Catalog No. 19035370 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Product Type:
PowerHeads™ Bar
Catalog No. Product Type
19-035-370 PowerHeads™ Bar
1 options
Catalog No. 19-035-370 Supplier Micronova™ Supplier No. PHB14
Specifications
Product Type PowerHeads™ Bar
Autoclavable Autoclavable
Width (English) 14 in.
For Use With (Equipment) QD-PHH Powerhead Handles
For Use With (Application) For cleaning in cleanrooms, pharmaceutical environments, research and animal labs
Material Stainless Steel