Learn More
ZOLL™ Powerheart™ G5 AED Defibrillation Pads- ICPR Pads
Catalog No. 17478087 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Pads, ICPR
Catalog No. Product Type
17-478-087 Pads, ICPR
1 options
This item is not returnable. View return policy
Catalog No. 17-478-087 Supplier ZOLL™ Supplier No. XELAED002B
This item is not returnable. View return policy
Non-polarized adult pads
  • In accordance with 2015 AHA Resuscitation Guidelines
Compatible With:
  • Powerheart™ G5
Notes:
  • Use of after-market defibrillation pads for Powerheart™ G5 AED voids AED warranty
Specifications
Product Type Pads, ICPR
For Use With (Equipment) Powerheart™ G5 AED