Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ Powerheart™ G5 AED Hard Sided Case

Catalog No. 17478091 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Case, Water Resistant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-478-091 Case, Water Resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-478-091 Supplier ZOLL™ Supplier No. XCAAED003A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water-resistant case for easy transport with reliable protection

  • Offers protection from bumps, drops and the elements
  • Interior foam for holding equipment
  • Can be locked

Holds:

  • Powerheart™ G5 AED
  • Ready kit
  • Extra set of pads

Includes:

  • Case
  • Two keys

Specifications

Product Type Case, Water Resistant
Color Black
Includes Two keys
For Use With (Equipment) Powerheart™ G5 AED, Ready kit, Extra set of pads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.