Learn More
ZOLL™ Powerheart™ G5 AED Pediatric Defibrillation Pads
Catalog No. 17478088 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Pads, Defibrillation
Catalog No. Product Type
17-478-088 Pads, Defibrillation
1 options
This item is not returnable. View return policy
Catalog No. 17-478-088 Supplier ZOLL™ Supplier No. XELAED003A
This item is not returnable. View return policy
Recommended for patients ages eight and under or those under 55 pounds
  • Non-polarized, can be placed on either position on chest and back
Compatible With:
  • Powerheart™ G5
Notes:
  • Use of after-market defibrillation pads voids AED warranty
Specifications
Product Type Pads, Defibrillation
Size Pediatric
For Use With (Application) For reducing confusion and saving time during a rescue
For Use With (Equipment) Powerheart™ G5 AED