Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ Powerheart™ G5 AED Premium Carry Case

Catalog No. 17478092 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Carry Case, G5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-478-092 Carry Case, G5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-478-092 Supplier ZOLL™ Supplier No. XCAAED007A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Semi-rigid case with adjustable shoulder strap for protection

  • D-ring on side of case for attaching Universal ready kit
  • Back pocket for extra storage of additional pads
  • Designed to keep Rescue Ready indicator and pads' expiration date visible

Compatible With:

  • Powerheart™ G5

Specifications

Product Type Carry Case, G5
Color Black
Includes D-ring
For Use With (Application) Protection and easy transport for rescue equipment
For Use With (Equipment) Powerheart™ G5 AED
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.