Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ Powerheart™ G5 Data USB Cable (A-to-A)

Catalog No. 17478094 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
USB Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-478-094 USB Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-478-094 Supplier ZOLL™ Supplier No. 500156801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For data exchange and configuring Powerheart™ G5

  • USB cable for Powerheart™ G5

Specifications

Product Type USB Cable
Color White
For Use With (Equipment) Powerheart™ G5 AED
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.