Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ Powerheart™ G5 Semi-Automatic AED

Catalog No. 17478085 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
AED Semi Auto Defibrillator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-478-085 AED Semi Auto Defibrillator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-478-085 Supplier ZOLL™ Supplier No. G5S80CS
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Displays rescue prompts and info with CPR feedback

  • Lithium battery
  • Cycle time: 15 seconds or less
  • Electrodes: Intellisense™ defibrillation pads
  • Information displayed: Rescue prompts and critical rescue info with CPR feedback corrective text prompts

Includes:

  • Soft carrying case
  • Universal ready kit with electrodes

Specifications

Product Type AED Semi Auto Defibrillator
Charging Time Up to 8 sec.
Energy Range 22 to 82 J (Pediatric), 95 to 354 J (Adult)
Includes Soft carrying case, Universal ready kit with electrodes
Operating Cycles 420 cycles/charge
Certifications/Compliance FDA approved
Height (English) 11.8 in.
Width (English) 9 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.