Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ Powerhouse™ 5 Channel Straight Cable Protectors

Catalog No. 19003161 Shop All Wearwell Tennessee Mat Company Inc. Products
Change view
Click to view available options
Style:
General Purpose
Heavy Duty
Medium Duty
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19-003-161 General Purpose
19-003-163 Medium Duty
19-003-162 Heavy Duty
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-003-161 Supplier Wearwell™ Supplier No. PWR.5CHAN.GP.BYL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Secure Lid-Lock feature protects pedestrian traffic by preventing trip hazards. Bright yellow lid is highly visible for enhanced safety and quick recognition.

  • Highly resistant to corrosion from oils and chemicals
  • Guards against impact and abrasion
  • Hidden hinges allow the lid to lie flat while protecting cables from damage
  • Hidden interlocks keep adjoining sections from lifting
  • Lid features a built-in access port for easy opening with a screwdriver or pry bar
  • Built-in underside handle makes relocating and handling easy
  • Secure Lid-Lock feature protects pedestrian traffic by preventing trip hazards
  • Available in General Purpose, Medium Duty, and Heavy Duty options

Specifications

Diameter (English) Cable Channel: 1.5 in.
Description General Purpose
Model PWR
Style General Purpose
Thickness (English) 3 in.
For Use With (Application) Cable Protection
Width (Metric) 12 cm
Color Black/Yellow
Dimensions (L x W) 91 x 12 cm
Length (Metric) 91 cm
Material Natural Rubber
Thickness (Metric) 7.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.