Learn More
Wearwell™ Powerhouse™ 5 Channel Straight Cable Protectors
Shop All Wearwell Tennessee Mat Company Inc. ProductsDescription
- Highly resistant to corrosion from oils and chemicals
- Guards against impact and abrasion
- Hidden hinges allow the lid to lie flat while protecting cables from damage
- Hidden interlocks keep adjoining sections from lifting
- Lid features a built-in access port for easy opening with a screwdriver or pry bar
- Built-in underside handle makes relocating and handling easy
- Secure Lid-Lock feature protects pedestrian traffic by preventing trip hazards
- Available in General Purpose, Medium Duty, and Heavy Duty options
Specifications
Specifications
| Model | PWR |
| Diameter (English) | Cable Channel: 1.25 in. |
| Color | Black/Yellow |
| Style | Medium Duty |
| Thickness (English) | 3 in. |
| Length (Metric) | 91 cm |
| Dimensions (L x W) | 91 x 12 cm |
| For Use With (Application) | Cable Protection |
| Thickness (Metric) | 7.6 cm |
| Material | Natural Rubber |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.