Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec PowerMop CE Battery

Catalog No. 17988188 Shop All Contec Inc. Products
Change view
Click to view available options
Product Type:
Replacement Lithium Ion Battery
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-988-188 Replacement Lithium Ion Battery
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-988-188 Supplier Contec Supplier No. PM400BATT
Add to Cart
Add to Cart

Replacement or additional battery for PowerMop CE.

  • Replacement or additional battery for PMCE 0400 PowerMop CE
  • PMCE 0400 PowerMop CE starter unit includes a battery and charger.
  • PowerMop CE is engineered for cleaning and residue removal, the dual counter-rotating pads deliver consistent, repeatable cleaning by effectively lifting particles, bioburden, and chemical residues from cleanroom floors.
  • ISO 14644-5 Certified for use in cleanroom environments.
  • HEPA filtration of the vacuum exhaust captures airborne particles during use.
  • Multi-layered cleaning pads offer uniform contact and will not damage floor surfaces.
  • Mechanical action of the counter-rotating pads effectively removes residues and particles from floors.
  • Counter rotating pads propel the unit forward, reducing operator effort.
  • Compatible with a range of disinfectants and solutions.
  • Adjustable solution delivery allows flexibility based on room size and soil load.
  • Assembled and disassembled tool-free supporting the frequent cleaning and changeover protocols.
  • The compact design makes it easy to store in smaller cleanrooms.

Specifications

Product Type Replacement Lithium Ion Battery
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Reusable
Quantity 1 Ea.
For Use With (Equipment) PowerMop CE
Color-coded No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.