Learn More
Bio Rad Laboratories POWERPAC POWER SUPPLY
Catalog No. NC9003275
Encompass
PowerPac Basic Power Supply , 100-120/220-240 V, includes power cord, instructions ,for basic applications such as submerged horizontal and mini vertical gel electrophoresis,provides simple programming in a compact, stackable case
Catalog No. NC9003275 Supplier Bio Rad Laboratories Supplier No. 1645050
PowerPac Basic Power Supply , 100-120/220-240 V, includes power cord, instructions ,for basic applications such as submerged horizontal and mini vertical gel electrophoresis,provides simple programming in a compact, stackable case