Learn More
MSC Guardair Wet/Dry Vacuum, Pneumatic, ATEX Certified, 55 gal, 1 ea
Catalog No. 502866714
Encompass
The Guardair PowerQUAD ATEX Vacuum Family provides pneumatic vacuum technology for exceptional performance. These ATEX-certified wet/dry vacuums are suitable for vacuuming chips, dust, debris, liquids, or sludge in various hazardous locations. Powered by compressed air, they are safe for wet, dry, or hazardous environments due to the absence of motors or electrical cords.
  • Pneumatic vacuum technology
  • ATEX-certified for hazardous locations
  • Electrically bonded components and multiple grounding options
  • Compressed air powered, no electric motors or cords
  • Suitable for wet, dry, or hazardous locations
  • Consumes up to 70% less compressed air
  • Equipped with a liquid shutoff assembly for wet and dry applications
  • Rugged, diecast aluminum, vacuum generating engine
  • Backwards compatible with existing pneumatic vacuum components
Catalog No. 50-286-6714 Supplier MSC Supplier No. 64359169
The Guardair PowerQUAD ATEX Vacuum Family provides pneumatic vacuum technology for exceptional performance. These ATEX-certified wet/dry vacuums are suitable for vacuuming chips, dust, debris, liquids, or sludge in various hazardous locations. Powered by compressed air, they are safe for wet, dry, or hazardous environments due to the absence of motors or electrical cords.
  • Pneumatic vacuum technology
  • ATEX-certified for hazardous locations
  • Electrically bonded components and multiple grounding options
  • Compressed air powered, no electric motors or cords
  • Suitable for wet, dry, or hazardous locations
  • Consumes up to 70% less compressed air
  • Equipped with a liquid shutoff assembly for wet and dry applications
  • Rugged, diecast aluminum, vacuum generating engine
  • Backwards compatible with existing pneumatic vacuum components