Learn More
MSC Guardair Wet/Dry Vacuum, 55 gal. ATEX Certified, 1 Kit
Catalog No. 502868925
Encompass
The PowerQUAD ATEX Vacuum Kit is a pneumatic, ATEX-certified vacuum engineered for hazardous locations, efficiently handling combustible dust, chips, liquids, or sludge. It features electrically bonded components and static conductive attachments to eliminate static electricity. Compressed air-powered, it operates without motors or electric cords, offering superior suction and significant operating cost savings.
  • ATEX certified for hazardous locations
  • Consumes up to 70% less compressed air
  • Customizable performance with three injector nozzle sets
  • Intrinsically safe; no electric motors, sparks, or cords
  • Rugged, diecast aluminum, 2-stage multi-venturi vacuum engine
  • Liquid shutoff assembly for wet and dry applications
  • Backwards compatible with existing pneumatic vacuum components
Catalog No. 50-286-8925 Supplier MSC Supplier No. 64360076
The PowerQUAD ATEX Vacuum Kit is a pneumatic, ATEX-certified vacuum engineered for hazardous locations, efficiently handling combustible dust, chips, liquids, or sludge. It features electrically bonded components and static conductive attachments to eliminate static electricity. Compressed air-powered, it operates without motors or electric cords, offering superior suction and significant operating cost savings.
  • ATEX certified for hazardous locations
  • Consumes up to 70% less compressed air
  • Customizable performance with three injector nozzle sets
  • Intrinsically safe; no electric motors, sparks, or cords
  • Rugged, diecast aluminum, 2-stage multi-venturi vacuum engine
  • Liquid shutoff assembly for wet and dry applications
  • Backwards compatible with existing pneumatic vacuum components