Learn More
MSC Guardair Vacuum Kit, ATEX, 30 Gallon, 1.5 inch Inlet
Catalog No. 502836886
Encompass
This pneumatic vacuum kit is designed for high performance and safety in hazardous environments. ATEX-certified, it is suitable for vacuuming combustible dust, liquids, or sludge and operates in various hazardous locations. The kit includes static conductive hoses and metal accessories.
  • ATEX certified for hazardous locations.
  • Energy efficient and powerful, consuming up to 70% less compressed air.
  • Customizable performance with injector nozzle sets.
  • Intrinsically safe, preventing electrical sparks and using no electric cords.
  • Proprietary 2-stage, multi-venturi engine design for superior performance.
  • Versatile, with liquid shutoff assembly for wet/dry applications.
  • Compatible with existing pneumatic vacuum components.
Catalog No. 50-283-6886 Supplier MSC Supplier No. 64357858
This pneumatic vacuum kit is designed for high performance and safety in hazardous environments. ATEX-certified, it is suitable for vacuuming combustible dust, liquids, or sludge and operates in various hazardous locations. The kit includes static conductive hoses and metal accessories.
  • ATEX certified for hazardous locations.
  • Energy efficient and powerful, consuming up to 70% less compressed air.
  • Customizable performance with injector nozzle sets.
  • Intrinsically safe, preventing electrical sparks and using no electric cords.
  • Proprietary 2-stage, multi-venturi engine design for superior performance.
  • Versatile, with liquid shutoff assembly for wet/dry applications.
  • Compatible with existing pneumatic vacuum components.