Learn More
MSC Guardair PowerQUAD Maintenance Kit, 5/10 Gallon
Catalog No. 502837602
Encompass
The PowerQUAD 5/10 Gallon Maintenance Kit is designed to rebuild worn parts in PowerQUAD 5/10 Gallon Vacuums. It includes a comprehensive set of replacement components to ensure the continued optimal performance of the vacuum system.
  • Rebuilds worn parts
  • Includes (1) exhaust silencer foam inserts
  • Includes (1) cover gasket
  • Includes (1) standard cartridge filter
  • Includes (1) second stage shutoff gasket
  • Includes (1) vacuum engine gasket
  • Includes (1) 5/10 gal wire cage coalescing filter
  • Ensures proper maintenance of 5/10 gallon vacuums
  • Extends the lifespan of the vacuum by replacing critical worn components
Catalog No. 50-283-7602 Supplier MSC Supplier No. 64362387
The PowerQUAD 5/10 Gallon Maintenance Kit is designed to rebuild worn parts in PowerQUAD 5/10 Gallon Vacuums. It includes a comprehensive set of replacement components to ensure the continued optimal performance of the vacuum system.
  • Rebuilds worn parts
  • Includes (1) exhaust silencer foam inserts
  • Includes (1) cover gasket
  • Includes (1) standard cartridge filter
  • Includes (1) second stage shutoff gasket
  • Includes (1) vacuum engine gasket
  • Includes (1) 5/10 gal wire cage coalescing filter
  • Ensures proper maintenance of 5/10 gallon vacuums
  • Extends the lifespan of the vacuum by replacing critical worn components