Learn More
MSC Guardair Vacuum Kit, 55 Gallon, NED, 2 in Inlet
Catalog No. 502835621
Encompass
This pneumatic vacuum kit efficiently cleans up chips, dust, debris, liquids, or sludge. It includes electrically bonded components and static conductive attachments to eliminate static electricity. Powered by compressed air, it operates without motors or electric cords, offering superior vacuum suction and operating cost savings.
  • Consumes up to 70% less compressed air with high vacuum suction and cost savings.
  • Operates quietly at 88 dBA or less.
  • Customizable performance with three injector nozzle sets.
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
  • Equipped with a liquid shutoff for wet and dry applications.
Catalog No. 50-283-5621 Supplier MSC Supplier No. 50322866
This pneumatic vacuum kit efficiently cleans up chips, dust, debris, liquids, or sludge. It includes electrically bonded components and static conductive attachments to eliminate static electricity. Powered by compressed air, it operates without motors or electric cords, offering superior vacuum suction and operating cost savings.
  • Consumes up to 70% less compressed air with high vacuum suction and cost savings.
  • Operates quietly at 88 dBA or less.
  • Customizable performance with three injector nozzle sets.
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
  • Equipped with a liquid shutoff for wet and dry applications.