Learn More
MSC Guardair Vacuum Kit, PowerQUAD NED, 30 Gallon, 1.5 in Inlet
Catalog No. 502835973
Encompass
The PowerQUAD 30-Gallon NED Vacuum Kit with a 1.5" Inlet is a pneumatic vacuum designed for wet and dry industrial applications like chips, dust, debris, or sludge. It utilizes electrically bonded components and static-conductive attachments to eliminate static electricity. Powered by compressed air, it avoids rotating parts or electric cords, offering superior suction and operating cost savings.
  • Consumes up to 70% less compressed air for powerful vacuum suction.
  • Operates quietly at 88 dBA or less.
  • Customizable performance via three color-coded, easy-to-change injector nozzle sets.
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi vacuum generating engine.
  • Equipped with a liquid shutoff assembly, suitable for both wet and dry applications.
Catalog No. 50-283-5973 Supplier MSC Supplier No. 64357957
The PowerQUAD 30-Gallon NED Vacuum Kit with a 1.5" Inlet is a pneumatic vacuum designed for wet and dry industrial applications like chips, dust, debris, or sludge. It utilizes electrically bonded components and static-conductive attachments to eliminate static electricity. Powered by compressed air, it avoids rotating parts or electric cords, offering superior suction and operating cost savings.
  • Consumes up to 70% less compressed air for powerful vacuum suction.
  • Operates quietly at 88 dBA or less.
  • Customizable performance via three color-coded, easy-to-change injector nozzle sets.
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi vacuum generating engine.
  • Equipped with a liquid shutoff assembly, suitable for both wet and dry applications.