Learn More
MSC Guardair Wet/Dry Vacuum, 55 gal Capacity, Air Powered, 2 in Inlet, Steel Tank
Catalog No. 502837276
Encompass
This 55 gal NED vacuum kit, powered by compressed air, delivers exceptional performance for vacuuming chips, dust, debris, liquids, or sludge. It features electrically bonded and static conductive components to eliminate static electricity. Operating without motors or electric cords, it provides superior suction with significant cost savings.
  • Consumes up to 70% less compressed air for high suction and cost savings.
  • Quiet operation at 88 dBA or less.
  • Customizable performance via three injector nozzle sets.
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
  • Liquid shutoff assembly supports wet and dry applications.
  • Electrically bonded and static conductive components eliminate static electricity.
  • No rotating parts, motors, or electric cords, enhancing safety.
Catalog No. 50-283-7276 Supplier MSC Supplier No. 64360241
This 55 gal NED vacuum kit, powered by compressed air, delivers exceptional performance for vacuuming chips, dust, debris, liquids, or sludge. It features electrically bonded and static conductive components to eliminate static electricity. Operating without motors or electric cords, it provides superior suction with significant cost savings.
  • Consumes up to 70% less compressed air for high suction and cost savings.
  • Quiet operation at 88 dBA or less.
  • Customizable performance via three injector nozzle sets.
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
  • Liquid shutoff assembly supports wet and dry applications.
  • Electrically bonded and static conductive components eliminate static electricity.
  • No rotating parts, motors, or electric cords, enhancing safety.