Learn More
MSC Guardair Vacuum Kit, 55 Gallon, PulseAir Technology, 2 inch Inlet
Catalog No. 502836490
Encompass
This pneumatic vacuum kit is designed for high-performance clean-up in high dust environments. It features PulseAir technology for on-demand filter cleaning, reducing dust exposure and meeting OSHA Silica Dust Regulation 29 CFR 1926.1153. Electrically bonded components manage static electricity. Operating on compressed air, it avoids electrical hazards, generates superior vacuum suction, and offers substantial operating cost savings.
  • Energy efficient, consumes up to 70% less compressed air.
  • Quiet operation at 88 dBA or less.
  • Customizable performance with three injector nozzle sets.
  • Proprietary 2-stage, multi-venturi engine for superior suction.
  • Kit includes static conductive vacuum and air supply hoses, HEPA filter, and tools.
  • Backed by a Forever Warranty.
Catalog No. 50-283-6490 Supplier MSC Supplier No. 64360217
This pneumatic vacuum kit is designed for high-performance clean-up in high dust environments. It features PulseAir technology for on-demand filter cleaning, reducing dust exposure and meeting OSHA Silica Dust Regulation 29 CFR 1926.1153. Electrically bonded components manage static electricity. Operating on compressed air, it avoids electrical hazards, generates superior vacuum suction, and offers substantial operating cost savings.
  • Energy efficient, consumes up to 70% less compressed air.
  • Quiet operation at 88 dBA or less.
  • Customizable performance with three injector nozzle sets.
  • Proprietary 2-stage, multi-venturi engine for superior suction.
  • Kit includes static conductive vacuum and air supply hoses, HEPA filter, and tools.
  • Backed by a Forever Warranty.