Learn More
MSC Guardair Wet/Dry Vacuum, Air, 20 Gallon, HEPA Filtration
Catalog No. 502838453
Encompass
This Guardair PowerQUAD PulseAir Wet/Dry Vacuum is engineered for exceptional performance in high dust environments. It features HEPA, cartridge, and coalescing filters within a robust steel tank, utilizing pneumatic technology for safety and efficiency.
  • Advanced HEPA, cartridge, and coalescing filtration system for superior air purification.
  • PulseAir technology allows on-demand filter cleaning without opening the unit, meeting OSHA silica dust regulation.
  • Pneumatic power ensures safety by eliminating motors, sparks, and electrical cords.
  • Highly efficient operation consumes up to 70% less compressed air while maintaining strong suction.
  • Quiet operation at 88 dBA or less.
  • Robust diecast aluminum engine with 2-stage, multi-venturi design.
  • Suitable for both wet and dry applications.
  • Static dissipative design eliminates static electricity.
Catalog No. 50-283-8453 Supplier MSC Supplier No. 64357635
This Guardair PowerQUAD PulseAir Wet/Dry Vacuum is engineered for exceptional performance in high dust environments. It features HEPA, cartridge, and coalescing filters within a robust steel tank, utilizing pneumatic technology for safety and efficiency.
  • Advanced HEPA, cartridge, and coalescing filtration system for superior air purification.
  • PulseAir technology allows on-demand filter cleaning without opening the unit, meeting OSHA silica dust regulation.
  • Pneumatic power ensures safety by eliminating motors, sparks, and electrical cords.
  • Highly efficient operation consumes up to 70% less compressed air while maintaining strong suction.
  • Quiet operation at 88 dBA or less.
  • Robust diecast aluminum engine with 2-stage, multi-venturi design.
  • Suitable for both wet and dry applications.
  • Static dissipative design eliminates static electricity.