Learn More
MSC Guardair ULPA Filter, 0.1 Micron, 99.999% Efficiency, Microglass Media, with Pre-Filter Bag
Catalog No. 502836495
Encompass
The PowerQUAD ULPA (Ultra Low Penetration Air) Cartridge Filter is engineered to capture minute particles, meeting high industry standards for air filtration. It offers 0.1 micron filtration at a minimum of 99.999% efficiency, ideal for environments with allergens, mold, pollen, and superfine powders. A Cloth Pre-Filter Bag is included, providing extra filtration and extending the cartridge filter's life.
  • Filtration media: Micro glass.
  • 0.1 micron filtration with 99.999% minimum efficiency.
  • Constructed with a galvanized carbon steel cage and end cap.
  • Includes a napped cotton flannel pre-filter bag for extended filter life.
  • Compatible with 55, 30, and 20 gallon drum vacuums and vacuum generating heads.
  • Ensures clean air quality and is effective against allergens, mold, pollen, and superfine powders.
Catalog No. 50-283-6495 Supplier MSC Supplier No. 64360894
The PowerQUAD ULPA (Ultra Low Penetration Air) Cartridge Filter is engineered to capture minute particles, meeting high industry standards for air filtration. It offers 0.1 micron filtration at a minimum of 99.999% efficiency, ideal for environments with allergens, mold, pollen, and superfine powders. A Cloth Pre-Filter Bag is included, providing extra filtration and extending the cartridge filter's life.
  • Filtration media: Micro glass.
  • 0.1 micron filtration with 99.999% minimum efficiency.
  • Constructed with a galvanized carbon steel cage and end cap.
  • Includes a napped cotton flannel pre-filter bag for extended filter life.
  • Compatible with 55, 30, and 20 gallon drum vacuums and vacuum generating heads.
  • Ensures clean air quality and is effective against allergens, mold, pollen, and superfine powders.